1-Boc-2-methylpiperazine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylpiperazine, N1-BOC protected, tert-Butyl 2-methylpiperazine-1-carboxylate

A154682-10G A154682-1G A154682-25G A154682-5G
103804-670EA 9 USD
103804-670 103842-862 76689-500 77300-070 77318-600 77318-602
1-Boc-2-methylpiperazine 97%
1-Boc-2-methylpiperazine

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 268.7 °C (760 mm Hg)
Flash Pt: 116.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179168
CAS Number: 120737-78-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR