tert-Butyl 2-methylhydrazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-methylhydrazinecarboxylate

A238888-100MG A238888-250MG A238888-1G
103811-254EA 15 USD
103811-254 103851-050 76815-342 76815-344 77090-188
tert-Butyl 2-methylhydrazine-1-carboxylate 98%
tert-Butyl 2-methylhydrazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₆H₁₄N₂O₂
MW: 146.188 g/mol
Melting Pt: 49…51 °C
Storage Temperature: Freezer
MDL Number: MFCD16660139
CAS Number: 127799-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR