tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Methylpiperidin-4-one, N-BOC protected, tert-Butyl 2-methyl-4-oxopiperidine-1-carboxylate
A273733-1G
A273733-25G
A273733-10G
103813-154EA
21
USD
103813-154
103853-078
77305-142
77305-144
77593-690
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate 97%
tert-Butyl-2-methyl-4-oxopiperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₃ MW: 213.2735 g/mol Boiling Pt: 298.7 °C (760 mm Hg) Melting Pt: 48…51 °C Flash Pt: 134.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04035608 CAS Number: 190906-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...