tert-Butyl glycolate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-hydroxyacetate

A200188-5G A200188-250MG A200188-1G A200188-500G A200188-10G
103817-172EA 6 USD
103817-172 103847-800 77085-862 77274-788 77302-658 77550-276 77593-296
tert-Butyl glycolate 98%
tert-Butyl glycolate

Precaución: for Research Use Only

Formula: C₆H₁₂O₃
MW: 132.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03452739
CAS Number: 50595-15-8
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR