tert-Butyl-2-hydroxy-2-methylpropylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-hydroxy-2-methylpropylcarbamate

A222680-250MG A222680-1G
103818-992EA 24 USD
103818-992 103849-870 76975-876 77088-484
tert-Butyl-2-hydroxy-2-methylpropylcarbamate 98%
tert-Butyl-2-hydroxy-2-methylpropylcarbamate

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.2500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09608009
CAS Number: 183059-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR