tert-Butyl-2-formylpiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-formylpiperidine-1-carboxylate, 1-tert-Butoxycarbonylpiperidine-2-carboxaldehyde

A218570-10G A218570-1G A218570-5G
103818-718EA 12 USD
103818-718 103849-588 77013-208 77088-112 77088-114
tert-Butyl-2-formylpiperidine-1-carboxylate 95%
tert-Butyl-2-formylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.2700 g/mol
Boiling Pt: 252.8 °C (760 mm Hg)
Melting Pt: 96…97 °C
Flash Pt: 132.5 °C
Storage Temperature: Freezer
MDL Number: MFCD02179021
CAS Number: 157634-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR