tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate 95%

Supplier: Ambeed
Warning

Synonyms: 4-(tert-Butoxycarbonylamino)-3-fluorophenylboronic acid, pinacol ester

A190537-1G A190537-5G A190537-250MG
103816-304EA 27 USD
103816-304 103846-906 77084-712 77084-714 77097-130
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate 95%
tert-Butyl 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenylcarbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₅BFNO₄
MW: 337.194 g/mol
Boiling Pt: 376.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09033883
CAS Number: 262444-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR