1-Boc-2-cyanopiperidine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-cyanopiperidine-1-carboxylate
A323836-25G
A323836-5G
A323836-1G
103825-360EA
9
USD
103825-360
103836-302
76839-218
76839-220
77306-314
1-Boc-2-cyanopiperidine 97%
1-Boc-2-cyanopiperidine
Precaución: for Research Use Only
Formula:
C₁₁H₁₈N₂O₂ MW: 210.27 g/mol Boiling Pt: 325.3 °C (760 mm Hg) Melting Pt: 60…64 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01861221 CAS Number: 153749-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...