tert-Butyl cyanoacetate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-cyanoacetate

A267483-100G A267483-10G A267483-500G A267483-25G A267483-5G
103852-754EA 5 USD
103852-754 76787-120 76787-122 76787-124 77092-494 77092-496
tert-Butyl cyanoacetate 98%
tert-Butyl cyanoacetate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂
MW: 141.17 g/mol
Boiling Pt: 194.6 °C (760 mm Hg)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001938
CAS Number: 1116-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR