tert-Butyl-2-chloropyridine-4-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-chloroisonicotinate
A407962-250MG
A407962-1G
A407962-25G
A407962-5G
103827-172EA
8
USD
103827-172
103838-214
76694-814
76841-712
76841-714
76841-716
tert-Butyl-2-chloropyridine-4-carboxylate 97%
tert-Butyl-2-chloropyridine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂ClNO₂ MW: 213.67 g/mol Boiling Pt: 287.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD03411667 CAS Number: 295349-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...