tert-Butyl 2-carbamoylpyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-carbamoyllpyrrolidine-1-carboxylate

A275240-100G A275240-10G A275240-250MG A275240-1G A275240-25G A275240-5G
103813-242EA 6 USD
103813-242 103853-168 76787-274 76787-276 76787-278 77093-012 77093-014 77093-016
tert-Butyl 2-carbamoylpyrrolidine-1-carboxylate 95%
tert-Butyl 2-carbamoylpyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.26 g/mol
Boiling Pt: 370.1 °C (760 mm Hg)
Flash Pt: 177.6 °C
Storage Temperature: Ambient
MDL Number: MFCD02183583
CAS Number: 54503-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR