tert-Butyl-2-aminopropylcarbamate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2-aminopropylcarbamate
A238203-10G
A238203-250MG
A238203-5G
103811-206EA
106
USD
103811-206
103850-990
76976-378
76976-380
77090-102
tert-Butyl-2-aminopropylcarbamate 98%
tert-Butyl-2-aminopropylcarbamate
Precaución: for Research Use Only
Formula:
C₈H₁₈N₂O₂ MW: 174.2407 g/mol Boiling Pt: 263.7 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09788609 CAS Number: 255735-88-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...