Glycine-tert-butyl ester(H-Gly-OtBu) 97%

Supplier: Ambeed
Warning

A151947-10G A151947-1G A151947-25G A151947-5G A151947-100G A151947-500G
103804-322EA 8 USD
103804-322 103842-472 76812-240 77318-052 77318-054 77318-056 77567-192 77671-128
Glycine-tert-butyl ester(H-Gly-OtBu) 97%

Precaución: For research use only.

Formula: C₆H₁₃NO₂
Boiling Pt: 145.7 °C (760 mm Hg)
Melting Pt: 29…31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038194
CAS Number: 6456-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR