2-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-amino-7-azaspiro[3.5]nonane-7-carboxylate

A289595-100MG A289595-250MG A289595-1G A289595-5G A289595-25G
103813-840EA 24 USD
103813-840 103834-980 76816-536 76816-538 76837-562 76837-564 77672-488
2-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester 95%
2-Amino-7-azaspiro[3.5]nonane-7-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
Boiling Pt: 336.5 °C (760 mm Hg)
Flash Pt: 157.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14581153
CAS Number: 1239319-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR