H-Aib-OtBu·HCl 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-aminoisobutyrate hydrochloride, tert-Butyl 2-amino-2-methylpropanoate hydrochloride

A411441-10G A411441-1G A411441-100G
103827-226EA 5 USD
103827-226 103838-276 76817-974 76841-816 77673-000
H-Aib-OtBu·HCl 95%
H-Aib-OtBu·HCl

Precaución: for Research Use Only

Formula: C₈H₁₇NO₂·HCl
MW: 195.7 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08272284
CAS Number: 84758-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR