tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate 95%

Supplier: Ambeed
Warning

A422550-1G A422550-100MG A422550-250MG
103827-420EA 44 USD
103827-420 103838-468 76842-092 77168-546 77168-548
tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate 95%
tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
Boiling Pt: 337 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06739056
CAS Number: 336191-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR