tert-Butyl-2,7-diazaspiro[4.4]nonane-2-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A246466-5G
A246466-250MG
A246466-1G
103811-714EA
25
USD
103811-714
103851-548
77090-840
77165-898
77165-900
77304-392
tert-Butyl-2,7-diazaspiro[4.4]nonane-2-carboxylate 97%
tert-Butyl-2,7-diazaspiro[4.4]nonane-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₂N₂O₂ Boiling Pt: 319.4 °C (760 mm Hg) Flash Pt: 147 °C Storage Temperature: Freezer |
MDL Number:
MFCD03790957 CAS Number: 236406-49-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...