tert-Butyl-2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride

A224877-100MG A224877-250MG A224877-1G A224877-5G A224877-10G
103819-140EA 17 USD
103819-140 103850-032 76786-108 76786-110 77303-722 77303-724 77568-176
tert-Butyl-2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride 97%
tert-Butyl-2,7-diazaspiro[3.5]nonane-7-carboxylate hydrochloride

Precaución: For research use only.

Formula: C₁₂H₂₃ClN₂O₂
MW: 262.7762 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13185086
CAS Number: 1023301-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR