2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%

Supplier: Ambeed
Warning

A185432-250MG A185432-10G A185432-25G
103815-852EA 146 USD
103815-852 103846-418 76974-600 77881-395 77881-396
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride 95%
2,7-Diazaspiro[3.5]nonane-2-carboxylicacid, 1,1-dimethylethylester, hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂·ClH
Storage Temperature: Refrigerator
MDL Number: MFCD09835396
CAS Number: 929302-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR