2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate(2:1)

A318827-1G A318827-100G A318827-10G A318827-250MG A318827-25G A318827-5G
103825-186EA 11 USD
103825-186 103836-120 76838-972 76989-084 76989-086 76989-088 77306-190 77306-192
2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate 97%
2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate

Precaución: for Research Use Only

Formula: C₂₂H₃₈N₄O₈
MW: 486.57 g/mol
Melting Pt: 206…209 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12404932
CAS Number: 1041026-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR