2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester 97%

Supplier: Ambeed
Warning

A254304-1G A254304-5G A254304-100MG
103812-128EA 38 USD
103812-128 103851-982 77091-422 77091-424 77250-006
2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester 97%
2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₂
Boiling Pt: 276 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411671
CAS Number: 198989-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR