tert-Butyl-2,4-dichloro-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 2,4-dichloro-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate
A252912-100MG
A252912-10G
A252912-250MG
A252912-1G
A252912-25G
A252912-5G
103812-060EA
20
USD
103812-060
103851-908
77014-028
77014-030
77014-032
77091-334
77304-590
77304-592
tert-Butyl-2,4-dichloro-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate 98%
tert-Butyl-2,4-dichloro-5H-pyrrolo[3,4-d]pyrimidine-6(7H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₃Cl₂N₃O₂ MW: 290.1500 g/mol Boiling Pt: 405.8 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD08273919 CAS Number: 903129-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...