tert-Butyl-2,2-dimethylpiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,2-dimethylpiperazine-1-carboxylate

A140497-100MG A140497-10G A140497-250MG
103802-876EA 20 USD
103802-876 76984-152 76984-154 77592-668
tert-Butyl-2,2-dimethylpiperazine-1-carboxylate 98%
tert-Butyl-2,2-dimethylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
Boiling Pt: 275.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07371652
CAS Number: 674792-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR