N'-(Oxan-4-yl)(tert-butoxy)carbohydrazide 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(tetrahydro-2H-pyran-4-yl)hydrazinecarboxylate

A226179-100MG A226179-250MG
103819-230EA 54 USD
103819-230 103850-130 76874-120 76874-122
N'-(Oxan-4-yl)(tert-butoxy)carbohydrazide 97%
N'-(Oxan-4-yl)(tert-butoxy)carbohydrazide

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₃
MW: 216.277 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044978
CAS Number: 693287-79-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR