tert-Butyl (dimethylphosphono)acetate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(dimethoxyphosphoryl)acetate, Dimethylphosphonoacetic acid tert-Butyl ester, tert-Butyl dimethylphosphonoacetate, Dimethyl (tert-Butoxycarbonylmethyl)phosphonate, Dimethyl (Boc-methyl)phosphonate

A191394-100G A191394-10G A191394-1G A191394-25G A191394-5G
103846-970EA 6 USD
103846-970 76799-988 76799-990 76799-992 77084-806 77084-808
tert-Butyl (dimethylphosphono)acetate 95%
tert-Butyl (dimethylphosphono)acetate

Precaución: for Research Use Only

Formula: C₈H₁₇O₅P
MW: 224.1913 g/mol
Boiling Pt: 86…87 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00042939
CAS Number: 62327-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR