tert-Butyl-2-(bromomethyl)pyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 2-(bromomethyl)pyrrolidine-1-carboxylate

A123342-100MG A123342-250MG A123342-1G
103800-392EA 221 USD
103800-392 76971-672 76971-674 77312-508
tert-Butyl-2-(bromomethyl)pyrrolidine-1-carboxylate 95%
tert-Butyl-2-(bromomethyl)pyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₈BrNO₂
MW: 264.17 g/mol
Boiling Pt: 300.1±15.0 °C (760 mm Hg)
Flash Pt: 135.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425263
CAS Number: 181258-46-8
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR