tert-Butyl (4-oxopiperidin-1-yl)acetate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(4-oxopiperidin-1-yl)acetate

A513247-1G A513247-100MG A513247-250MG
103828-426EA 183 USD
103828-426 76843-514 77017-340 77017-342
tert-Butyl (4-oxopiperidin-1-yl)acetate 98%
tert-Butyl (4-oxopiperidin-1-yl)acetate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
MW: 213.273 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07371646
CAS Number: 149554-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR