tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate 97%

Supplier: Ambeed
Warning

Synonyms: 4-{2-[(tert-Butoxycarbonyl)amino]ethoxy}aniline, tert-Butyl 2-(4-aminophenoxy)ethylcarbamate

A102907-250MG A102907-1G A102907-5G
103796-446EA 36 USD
103796-446 103822-148 76780-576 77309-018 77309-020
tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate 97%
tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate

Precaución: for Research Use Only

Formula: C₁₃H₂₀N₂O₃
MW: 252.309 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11922406
CAS Number: 159184-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR