tert-Butyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate 97%

Supplier: Ambeed
Warning

A100474-100MG A100474-250MG A100474-1G
103795-462EA 99 USD
103795-462 103821-156 77008-276 77008-278 77296-704
tert-Butyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate 97%
tert-Butyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate

Precaución: for Research Use Only

Formula: C₁₅H₂₅BN₂O₄
MW: 308.18 g/mol
Boiling Pt: 407.4±25.0 °C (760 mm Hg)
Flash Pt: 200.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22570725
CAS Number: 1006875-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR