tert-Butyl 1-oxo-8-azaspiro[4.5]decane-8-carboxylate 97%
Supplier: Ambeed
|
Warning
|
A232914-1G
A232914-100MG
A232914-250MG
103810-864EA
29
USD
103810-864
103850-620
77089-564
77165-582
77165-584
tert-Butyl 1-oxo-8-azaspiro[4.5]decane-8-carboxylate 97%
tert-Butyl 1-oxo-8-azaspiro[4.5]decane-8-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₂₃NO₃ MW: 253.337 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18074085 CAS Number: 191805-29-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...