tert-Butyl-1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate, 2,8-Diazaspiro[4.5]decan-1-one, N8-BOC protected

A157177-100MG A157177-10G A157177-250MG A157177-5G
103804-978EA 16 USD
103804-978 103843-178 76812-504 76812-506 76812-508 77318-998
tert-Butyl-1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate 97%
tert-Butyl-1-oxo-2,8-diazaspiro[4.5]decane-8-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₃
MW: 254.33 g/mol
Boiling Pt: 424.9±38.0 °C (760 mm Hg)
Flash Pt: 210.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08461248
CAS Number: 268550-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR