tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%

Supplier: Ambeed
Warning

A184560-1G A184560-5G A184560-250MG A184560-100G
103815-758EA 6 USD
103815-758 103846-322 77083-940 77083-942 77236-654 77567-692
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate 97%
tert-Butyl-1-oxa-6-azaspiro[2.5]octane-6-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₉NO₃
Boiling Pt: 297.6 °C (760 mm Hg)
Flash Pt: 133.799 °C
Storage Temperature: Ambient
MDL Number: MFCD07779385
CAS Number: 147804-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR