tert-Butyl-1-oxa-5-azaspiro[2,5]octane-5-carboxylate 98%

Supplier: Ambeed
Warning

A213266-1G A213266-5G
103818-282EA 77 USD
103818-282 77237-432 77237-434
tert-Butyl-1-oxa-5-azaspiro[2,5]octane-5-carboxylate 98%
tert-Butyl-1-oxa-5-azaspiro[2,5]octane-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₃
Boiling Pt: 297.6 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD07779386
CAS Number: 276872-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR