N-tert-Butoxycarbonylimidazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl 1H-imidazole-1-carboxylate, N-Boc-imidazole
A179146-100G
A179146-500G
A179146-25G
A179146-5G
A179146-10G
103845-744EA
7
USD
103845-744
76974-386
76974-388
77083-196
77083-198
77671-534
N-tert-Butoxycarbonylimidazole 98%
N-tert-Butoxycarbonylimidazole
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O₂ MW: 168.20 g/mol Boiling Pt: 247.4 °C (760 mm Hg) Melting Pt: 45…47 °C Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00014497 CAS Number: 49761-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...