tert-Butyl-1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate

A134112-100MG A134112-250MG A134112-5G A134112-1G
103801-976EA 35 USD
103801-976 77027-204 77027-206 77027-208 77298-690
tert-Butyl-1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate 97%
tert-Butyl-1-bromo-5,6-dihydroimidazo[1,5-a]pyrazine-7(8H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₆BrN₃O₂
MW: 302.1700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13152218
CAS Number: 1188265-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR