tert-Butyl 1,6-diazaspiro[3.5]nonane-1-carboxylate 95%

Supplier: Ambeed
Warning

A197624-100MG A197624-250MG A197624-1G A197624-5G
103816-952EA 710 USD
103816-952 76825-462 76825-464 77302-562 77885-457
tert-Butyl 1,6-diazaspiro[3.5]nonane-1-carboxylate 95%
tert-Butyl 1,6-diazaspiro[3.5]nonane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₂N₂O₂
Boiling Pt: 326.1 °C (760 mm Hg)
Flash Pt: 180.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14581198
CAS Number: 1251005-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR