tert-Butyl-1,4-diazepane-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 1-BOC-hexahydro-1,4-diazepine, tert-Butyl 1,4-diazepane-1-carboxylate

A152823-100G A152823-10G A152823-1G A152823-25G A152823-5G
103804-436EA 6 USD
103804-436 103842-608 76689-420 76689-422 77318-238 77318-240 77318-242
tert-Butyl-1,4-diazepane-1-carboxylate 97%
tert-Butyl-1,4-diazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
Boiling Pt: 95…110 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00276987
CAS Number: 112275-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR