trans-4-(Boc-amino)cyclohexanol 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (trans-4-hydroxycyclohexyl)carbamate, trans-4-Aminocyclohexan-1-ol, N-BOC protected

A149616-100G A149616-10G A149616-1G A149616-500G A149616-25G
103842-180EA 6 USD
103842-180 76768-104 76768-106 76768-108 76768-110 77317-686
trans-4-(Boc-amino)cyclohexanol 95%
trans-4-(Boc-amino)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
Boiling Pt: 337.7 °C (760 mm Hg)
Melting Pt: 172…173 °C
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD03844613
CAS Number: 111300-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR