trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (trans-4-(hydroxymethyl)cyclohexyl)carbamate, tert-butyl N-[trans-4-(hydroxymethyl)cyclohexyl]carbamate

A133991-10G A133991-250MG A133991-1G A133991-25G A133991-5G A133991-100G
103801-948EA 6 USD
103801-948 76782-510 76782-512 77314-650 77314-652 77314-654 77599-926
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane 95%
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD03844605
CAS Number: 239074-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR