trans-tert-Butyl-N-(3-aminocyclobutyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (trans-3-aminocyclobutyl)carbamate, trans-Cyclobutane-1,3-diamine, N1-BOC protected

A482913-100MG A482913-250MG
103828-148EA 59 USD
103828-148 103839-222 76877-862 77613-512
trans-tert-Butyl-N-(3-aminocyclobutyl)carbamate 97%
trans-tert-Butyl-N-(3-aminocyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09040641
CAS Number: 871014-19-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR