tert-Butyl (trans-2-hydroxycyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

A135517-250MG A135517-1G A135517-5G
103802-166EA 13 USD
103802-166 76688-786 77298-788 77298-790
tert-Butyl (trans-2-hydroxycyclohexyl)carbamate 97%
tert-Butyl (trans-2-hydroxycyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD04974283
CAS Number: 121282-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR