tert-Butyl (trans-2-aminocyclohexyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (1R,2R)-2-aminocyclohexylcarbamate

A154160-250MG A154160-1G
103804-608EA 11 USD
103804-608 103842-788 76798-742 77318-482
tert-Butyl (trans-2-aminocyclohexyl)carbamate 95%
tert-Butyl (trans-2-aminocyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 322.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06795909
CAS Number: 137731-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR