N,O-Di-boc-hydroxylamine 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
N,O-Di-Boc-hydroxylamine, tert-Butyl N-(tert-Butoxycarbonyloxy)carbamate, N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl (tert-butoxycarbonyl)oxycarbamate, N-(tert-Butoxycarbonyloxy)carbamic acid tert-Butyl ester
A162893-1G
A162893-5G
103805-666EA
5
USD
103805-666
103843-924
77300-618
77319-998
N,O-Di-boc-hydroxylamine 97%
N,O-Di-boc-hydroxylamine
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₅ MW: 233.264 g/mol Melting Pt: 67…70 °C Storage Temperature: Freezer |
MDL Number:
MFCD00034797 CAS Number: 85006-25-3 UN: 3077 ADR: 9,Ⅲ |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...