tert-Butyl (piperidin-4-ylmethyl)carbamate 97%

Supplier: Ambeed
Warning

Synonyms: (4-Piperidylmethyl)carbamic acid tert-Butyl ester, tert-Butyl (4-Piperidylmethyl)carbamate, 4-(tert-Butoxycarbonylaminomethyl)piperidine, 4-(Boc-aminomethyl)piperidine

A141436-100G A141436-10G A141436-250MG A141436-1G A141436-5G A141436-500G
103802-996EA 14 USD
103802-996 76798-314 76798-316 76798-318 77316-156 77316-158 77667-052
tert-Butyl (piperidin-4-ylmethyl)carbamate 97%
tert-Butyl (piperidin-4-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
Boiling Pt: 321.8 °C (760 mm Hg)
Melting Pt: 106 °C
Flash Pt: 148.4 °C
Storage Temperature: Ambient
MDL Number: MFCD01631214
CAS Number: 135632-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR