tert-butyl (cis-4-hydroxycyclohexyl)carbamate 97%

Supplier: Ambeed
Warning

A311496-100G A311496-10G A311496-250MG A311496-1G A311496-25G A311496-5G
103814-666EA 6 USD
103814-666 103835-856 76787-920 76787-922 76787-924 76838-652 76838-654 76838-656
tert-butyl (cis-4-hydroxycyclohexyl)carbamate 97%
tert-butyl (cis-4-hydroxycyclohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₃
Boiling Pt: 337.7 °C (760 mm Hg)
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD15071470
CAS Number: 167081-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR