cis-(3-Boc-amino)-4-fluoropyrrolidine 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (cis-4-fluoropyrrolidin-3-yl)carbamate

A344962-1G A344962-100MG A344962-250MG
103825-912EA 164 USD
103825-912 103836-894 76839-946 76989-512 76989-514
cis-(3-Boc-amino)-4-fluoropyrrolidine 97%
cis-(3-Boc-amino)-4-fluoropyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₇FN₂O₂
MW: 204.25 g/mol
Boiling Pt: 295.3±40.0 °C (760 mm Hg)
Flash Pt: 132.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18791660
CAS Number: 169750-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR