tert-Butyl (azetidin-3-ylmethyl)carbamate hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Aminomethyl)azetidine hydrochloride, 3-BOC protected

A237669-250MG A237669-25G A237669-1G
103811-168EA 18 USD
103811-168 103850-950 76815-310 76815-312 77090-046
tert-Butyl (azetidin-3-ylmethyl)carbamate hydrochloride 97%
tert-Butyl (azetidin-3-ylmethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08752583
CAS Number: 1170108-38-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR