tert-Butyl (6-formylpyridin-2-yl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A135662-1G
A135662-100MG
A135662-250MG
103802-198EA
153
USD
103802-198
77298-806
77883-095
77883-096
tert-Butyl (6-formylpyridin-2-yl)carbamate 98%
tert-Butyl (6-formylpyridin-2-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄N₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD13188949 CAS Number: 956523-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...