tert-Butyl (6-ethynylpyridin-2-yl)carbamate 97%

Supplier: Ambeed
Warning

A214073-100MG A214073-250MG A214073-1G
103818-364EA 193 USD
103818-364 77261-160 77261-162 77303-324
tert-Butyl (6-ethynylpyridin-2-yl)carbamate 97%
tert-Butyl (6-ethynylpyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD29760421
CAS Number: 1934630-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR