(6-Chloro-4-iodopyridin-3-yl)carbamic acid tert-butyl ester 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (6-chloro-4-iodopyridin-3-yl)carbamate

A188441-100MG A188441-10G A188441-250MG A188441-25G A188441-5G A188441-1G
103816-100EA 30 USD
103816-100 103846-696 76825-168 76825-170 76825-172 77084-436 77084-438 77302-126
(6-Chloro-4-iodopyridin-3-yl)carbamic acid tert-butyl ester 98%
(6-Chloro-4-iodopyridin-3-yl)carbamic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClIN₂O₂
MW: 354.572 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977418
CAS Number: 400777-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR